Order Entry
United States
ContactUsLinkComponent
Calcium, Plasma Standard Solution, Ca 10000 µg/ml
Calcium, Plasma Standard Solution, Ca 10000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Calcium, Plasma Standard Solution, Ca 10000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00085314
  • UN No.:
    3264
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together