1-Butanol ≥99.0% for HPLC
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Boiling Pt:116…118 °C
Melting Pt:–90 °C
Density:0.81 g/mL
Flash Pt:35 °C (95 °F)
EINECS:200-751-6
UN:1120
ADR:3,III
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay:99.0% min.
Specification Test Results
Frequently Bought Together