Dichloromethane ≥99.7% stabilized for spectrophotometry
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Boiling Pt:39…40 °C
Melting Pt:–97 °C
Density:1.325 g/mL
EINECS:200-838-9
UN:1593
ADR:6.1,III
Specification Test Results
- Assay:99.7% min.
Specification Test Results
Frequently Bought Together