

Ethanol denatured, anhydrous for HPLC
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78 °C
Density:0.789 g/mL
Flash Pt:11 °C
UN:1987
ADR:3,II
:GHS02
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Frequently Bought Together