Magnesium, Plasma Standard Solution, Mg 10000 µg/ml
Supplier: BeanTown Chemical
Certificates
About this item
EC No: 231-714-2
Specifications
- MDL No.:MFCD00085308
- UN No.:3264
- GHS signal:Danger
Specifications
Frequently Bought Together
EC No: 231-714-2