Cerium(IV) sulfate standardised solution 0.1 N in aqueous solution
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:13590-82-4
:
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852
:332.24 g/mol
Density:1.05 g/ml
EINECS:237-029-5
UN:3264
ADR:8,III
Frequently Bought Together