Chloroform-[D₁] ≥99.8% (Isotopic) contains 0.03% TMS
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:865-49-6
:
Formula:CDCl₃
MDL Number:MFCD00000827
:120.38 g/mol
Boiling Pt:60.9 °C
Melting Pt:–64 °C
Density:1.5 g/mL
EINECS:212-742-4
UN:1888
ADR:6.1,III
Specification Test Results
- Assay:99.8%
Specification Test Results
Frequently Bought Together