Hafnium, Plasma Standard Solution, Hf 1,000 µg/ml
EC No: 231-595-7
Haz class: 8
Packing group: III
GHS symbol: GHS05
- MDL No.:MFCD00011032
- UN No.:1789
- GHS signal:Danger
Frequently Bought Together
EC No: 231-595-7
Haz class: 8
Packing group: III
GHS symbol: GHS05