Order Entry
United States
ContactUsLinkComponent
 
Potassium oleate 40% (w/w) in water, paste
Catalog # BT135535-1KG
Supplier:  BeanTown Chemical
CAS Number:  
Potassium oleate 40% (w/w) in water, paste
Catalog # BT135535-1KG
Supplier:  BeanTown Chemical
Supplier Number:  135535-1KG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    1 kg
  • Cat. No.:
    BT135535-1KG
  • Supplier no.:
    135535-1KG

Specifications

About this item

 :
 :320.56 g/mol
ADR:9,III
CAS Number:143-18-0
Formula:C₁₈H₃₃KO₂
MDL Number:MFCD00064243