Order Entry
United States
ContactUsLinkComponent
 
Nickel, Plasma Standard Solution, Ni 10000 µg/ml
 
Nickel, Plasma Standard Solution, Ni 10000 µg/ml

 

  • MDL No.:
    MFCD00011137
  • UN No.:
    3264
  • GHS signal:
    Danger

 

 

Frequently Bought Together