Dodecylbenzenesulfonic acid ≥70% (w/w) in isopropanol (IPA)
BT124395-500ML
:
- Size:500 ml
- Cat. No.:BT124395-500ML
- Supplier no.:124395-500ML
:
:326.5
:82 °C
:0.992 g/mL
:54 °C (129 °F)
:204-489-3
:2924
:8,III
:27176-87-0
:68584-22-5
:C₁₈H₃₀O₃S
:MFCD00066677