Xylene (mixture of isomers) ≥98.5% ACS
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Boiling Pt:137…140 °C
Melting Pt:<–25 °C
Density:0.86 g/mL
Flash Pt:25 °C (77 °F)
EINECS:215-535-7
UN:1307
ADR:3,III
Specification Test Results
- Assay:98.5% min.
- Color (APHA):≤10ppm
- Residue after evaporation:≤10ppm
- Specific gravity at 15.6°C:0.865-0.875
- Substances darkened by H2SO4:Passes test
- Paraxylene:Per lot
- Metaxylene:Per lot
- Orthoxylene:Per lot
- Total C9 aromatics:Per lot
- Benzene content:≤0.01%
- Toluene content:≤0.5%
- Non-aromatics:≤0.30%
- Ethyl benzene:≤25%
- Sulfur compounds:≤1ppm
- Water:≤0.05%
Specification Test Results
Frequently Bought Together