Borane tetrahydrofuran complex 1 M in THF stabilized
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:14044-65-6
:
Formula:C₄H₁₁BO
MDL Number:MFCD00012429
:85.94 g/mol
Density:0.898 g/mL
Flash Pt:–17 °C (1 °F)
EINECS:237-881-8
UN:3148
ADR:4.3,I
Frequently Bought Together