p-Xylene ≥99%
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:106-42-3
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556
:106.17 g/mol
Boiling Pt:137…138 °C
Melting Pt:12…13 °C
Density:0.861 g/mL
Flash Pt:25 °C (77 °F)
EINECS:203-396-5
UN:1307
ADR:3,III
Specification Test Results
- Assay:99%
Specification Test Results
Frequently Bought Together