Order Entry
United States
ContactUsLinkComponent
Hafnium, Plasma Standard Solution, Hf 10000 µg/ml
Hafnium, Plasma Standard Solution, Hf 10000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Hafnium, Plasma Standard Solution, Hf 10000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00011032
  • UN No.:
    1789
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together