Anti-AKR1D1 Polyclonal Antibody Pair
10536-038
: Abnova
- Antigen Name:Aldo-keto reductase family 1 member D1
- Antigen Symbol:AKR1D1
- Clonality:Polyclonal
- Host:Multiple
- Reactivity:Human
- Size:1 Set
- Gene ID:6718
- Antigen Synonyms:3o5bred|CBAS2|SRD5B1
- Cat. No.:10536-038
- Supplier no.:H00006718-AP22
This Set comes with matched Antibody Pair to detect and quantify protein level of human AKR1D1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AKR1D1 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-AKR1D1 (20 µg);
Type:
Antigen: AKR1D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human