Anti-MAPK13 Antibody Pair
Catalog # 10540-084
Supplier: Abnova
Specifications
- Antigen Name:Mitogen-Activated Protein Kinase 13
- Antigen Symbol:MAPK13
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:1 Set
- Gene ID:5603
- Antigen Synonyms:MAPK-13|MAPK 13|p38delta|PRKM13|SAPK4
- Cat. No.:10540-084
- Supplier no.:H00005603-PW2
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-MAPK13 (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-MAPK13 (50 µl)
Type:
Antigen: MAPK13
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human