Anti-SP110 Polyclonal Antibody Pair
Catalog # 10542-974
Supplier: Abnova
Specifications
- Antigen Name:SP110 Nuclear Body Protein
- Antigen Symbol:SP110
- Clonality:Polyclonal
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:1 Set
- Gene ID:3431
- Antigen Synonyms:VODI|IFI75|Speckled 110 kDa|IFI41|Sp110 nuclear body protein|IPR1
- Cat. No.:10542-974
- Supplier no.:H00003431-PW2
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-SP110 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-SP110 (50 µg)
Type:
Antigen: SP110
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human