Anti-CREB3 + CREB3L4 Antibody Pair
10537-580
: Abnova
- Antigen Symbol:CREB3 + CREB3L4
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:10488|148327
- Cat. No.:10537-580
- Supplier no.:DI0522
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CREB3 protein, and the other against the CREB3L4 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CREB3 Rabbit purified Polyclonal Antibody (20 µg) 2. CREB3L4 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CREB3 + CREB3L4
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: