Anti-CRK + FGFR1 Antibody Pair
Catalog # 10537-594
Supplier: Abnova
Specifications
- Antigen Symbol:CRK + FGFR1
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:1398|2260
- Cat. No.:10537-594
- Supplier no.:DI0350
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CRK protein, and the other against the FGFR1 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CRK Rabbit purified Polyclonal Antibody (20 µg) 2. FGFR1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CRK + FGFR1
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: