Anti-CRK + PDGFRA Antibody Pair
Catalog # 10537-600
Supplier: Abnova
Specifications
- Antigen Symbol:CRK + PDGFRA
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:1398|5156
- Cat. No.:10537-600
- Supplier no.:DI0310
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CRK protein, and the other against the PDGFRA protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CRK Rabbit purified Polyclonal Antibody (20 µg) 2. PDGFRA Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CRK + PDGFRA
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: