Anti-TP53 + MDM2 Antibody Pair
Catalog # 10543-586
Supplier: Abnova
Specifications
- Antigen Symbol:TP53 + MDM2
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:7157|4193
- Cat. No.:10543-586
- Supplier no.:DI0256
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the TP53 protein, and the other against the MDM2 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. TP53 Rabbit purified Polyclonal Antibody (20 µg) 2. MDM2 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: TP53 + MDM2
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: