Anti-CTNNA3 + CTNNB1 Antibody Pair
Catalog # 10537-682
Supplier: Abnova
Specifications
- Antigen Symbol:CTNNA3 + CTNNB1
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:29119|1499
- Cat. No.:10537-682
- Supplier no.:DI0114
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CTNNA3 protein, and the other against the CTNNB1 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CTNNA3 Rabbit purified Polyclonal Antibody (20 µg) 2. CTNNB1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CTNNA3 + CTNNB1
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: