Anti-CD19 + CD82 Antibody Pair
Catalog # 10537-050
Supplier: Abnova
Specifications
- Antigen Symbol:CD19 + CD82
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:930|3732
- Cat. No.:10537-050
- Supplier no.:DI0083
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CD19 protein, and the other against the CD82 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CD19 Rabbit purified Polyclonal Antibody (20 µg) 2. CD82 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CD19 + CD82
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype: IgG1
Reactivity: