FlexTech™ Gloves, Synthetic Seamless Knit with Nitrile Palm, Wells Lamont
A black, synthetic, seamless knit shell and black-foam nitrile coating compose these gloves.
- Abrasion and Puncture Resistance
- For Small-Parts Handling and Electronics Applications
- Black-Foam Nitrile Coating
- Seamless Knit Shell
They work in electronics and small-parts handling.
The gloves feature excellent dexterity, superior grip, and abrasion and puncture resistance.
: ANSI Cut Level 1.
: Gloves are sold by the dozen pairs.
- Catalog No:
- 89040-184
- 89040-186
- 89040-190
- 89040-192
- 89040-188
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner Material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Foam nitrile palm coating
- Foam nitrile palm coating
- Foam nitrile palm coating
- Foam nitrile palm coating
- Foam nitrile palm coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff Style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
Frequently Bought Together