Order Entry
United States
ContactUsLinkComponent
PALCAM Listeria Selective Supplement, acc. to van Netten et al., MilliporeSigma
PALCAM Listeria Selective Supplement, acc. to van Netten et al., MilliporeSigma
Catalog # EM1.12122.0010
Supplier:  MilliporeSigma
CAS Number:  
PALCAM Listeria Selective Supplement, acc. to van Netten et al., MilliporeSigma
Catalog # EM1.12122.0010
Supplier:  MilliporeSigma
Supplier Number:  1.12122.0010
CAS Number:  

Specifications

  • Description:
    PALCAM Listeria Selective Supplement
  • Size:
    10 x 1 vial
  • Incubation:
    Listeria 48 hrs. at 35 °C, aerobic; Other strains 72 hrs. at 30 °C, aerobic
  • Storage:
    Store at +2°C to +8°C.
  • Cat. No.:
    EM1.12122.0010
  • Supplier no.:
    1.12122.0010

Specifications

About this item

Supplement tested in PALCAM Listeria Selective agar (Base) acc

  • Greater than or equal to 50% recovery on test medium Listeria monocytogenes ATCC 13932 (WDCM 00021)
  • Greater than or equal to 50% recovery on test medium Listeria monocytogenes ATCC 19111 (WDCM 00020)
  • Greater than or equal to 50% recovery on test medium Listeria monocytogenes ATCC 35152 (WDCM 00109)
  • Greater than or equal to 50% recovery on test medium Listeria innocua ATCC 33090 (WDCM 00017)
  • Less than or equal to 0.01% recovery on test medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • Less than or equal to 0.01% recovery on test medium Enterococcus faecalis ATCC 19433 (WDCM 00009)
  • Less than or equal to 0.01% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Black zone Listeria monocytogenes ATCC 13932 (WDCM 00021) +
  • Black zone Listeria monocytogenes ATCC 19111 (WDCM 00020) +
  • Black zone Listeria monocytogenes ATCC 35152 (WDCM 00109) +
  • Black zone Listeria innocua ATCC 33090 (WDCM 00017) +

In North America 1.12122.0010 replaces and is identical to 1.12122.0017

to VAN NETTEN et al. art. no. 1.11755. Polymyxin B sulfate ( >= 10 % - < 30 % ) 1405-20-5 and acriflavine hydrochloride ( >= 10 % - < 30 % ) 69235-50-3.