Order Entry
United States
ContactUsLinkComponent
Pierce™ Amino Acid Calibration Standard for HPLC, Thermo Scientific
Pierce™ Amino Acid Calibration Standard for HPLC, Thermo Scientific
Catalog # PI20088
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Amino Acid Calibration Standard for HPLC, Thermo Scientific
Catalog # PI20088
Supplier:  Invitrogen
Supplier Number:  20088
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Amino Acid Standard H
  • Size:
    10×1 ml
  • Cat. No.:
    PI20088
  • Supplier no.:
    20088

Specifications

About this item

Thermo Scientific Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5 µMol/mL each in 0.1N HCl, for use as a high-purity calibration standard for HPLC analysis of protein hydrolysates.

  • Complete mixture—contains eighteen (18) amino acids for assessment of amino acid composition of peptide hydrolysates
  • Quantitative—contains exact amounts of each amino acid (2.5±0.1 µMol/mL, except cystine at 1.25±0.1 µMol/mL), enabling its use as a quantitation standard
  • Stable—dissolved in 0.1N hydrochloric acid and stabilized as 1 mL aliquots in sealed glass ampules
  • Standard H—designed for compositional amino acid analysis by HPLC methods of protein or peptide hydrolysates

This quantitative solution of eighteen purified amino acids in 0.1N HCl is an ideal reference standard for calibrating amino acid analyzers and assessing the composition of hydrolyzed peptides and proteins (hydrolysates). To permit standardization of microbiological and other assays, the mixture uses only the L-form of each amino acid. The molar concentration of each amino acid is verified by conventional amino acid analysis methods, making the mixture suitable as both a qualitative and quantitative standard. The solution is provided as stable 1 mL aliquots in sealed glass ampules.