Order Entry
United States
ContactUsLinkComponent
BAX® System X5 PCR Assay for Salmonella, Hygiena™, Qualicon Diagnostics LLC
BAX® System X5 PCR Assay for Salmonella, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10002-770
Supplier:  Hygiena
CAS Number:  
undefined
BAX® System X5 PCR Assay for Salmonella, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10002-770
Supplier:  Hygiena
Supplier Number:  KIT2025
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BAX System X5 PCR Assay for Salmonella
  • Time to Results:
    14-34 hours
  • Enrichment Time:
    9-29 hours
  • Sample Type:
    Meats, Poultry, Vegetables, Fruit juices, Dairy, Peanut butter and Environmental
  • Processing Time:
    3.5 hours
  • Cat. No.:
    10002-770
  • Target group:
    Salmonella spp.
  • Supplier no.:
    D15407187
  • No. of tests:
    64

Specifications

About this item

The BAX® System X5 PCR Assay for Salmonella detects the target organism from standard enrichments of a variety of food and environmental samples

Each BAX System X5 PCR assay includes all the reagents you need to process 64 enriched samples – enough for two full process runs in the BAX® System X5 instrument. BAX System X5 PCR tablets contain a built-in control and are pre-packaged in PCR tubes to eliminate the need for reagent preparation.

The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.

The method includes automated sample preparation in the DuPont™ thermal block, followed by automated amplification and detection in the BAX® System X5 instrument.

Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System X5 instrument.