Silicone antifoam emulsion 10% active compound, Dow Corning® DB 110 A
Catalog # 10139-070
Supplier: PerkinElmer
CAS Number:
Specifications
- Size:500 ml
- Cat. No.:10139-070
- Supplier no.:B0507226
Specifications
About this item
:
Storage Temperature:Ambient