ContactUsLinkComponent
Jomac® Extra Heavy Weight Terry Cloth Gloves, Loop Out, Knit Wrist, Wells Lamont
  80091-486
undefined
Jomac® Extra Heavy Weight Terry Cloth Gloves, Loop Out, Knit Wrist, Wells Lamont
  80091-486
 :  642HR
  • Glove color:
    White,
    Brown
  • Glove size:
    X-Large
  • Ambidextrous/Hand-specific:
    Ambidextrous
  • Cuff Style:
    Knit Wrist
  • Cat. No.:
    80091-486
  • Supplier no.:
    642HR
Jomac extra heavy weight terry cloth gloves resist heat up to 350 °F.
  • Ambidextrous
  • Washing machine friendly
  • Brown and white
The knit-wrist cuff keeps out dirt and debris.
Users can clean the ambidextrous brown and white gloves in the washing machine.
 : Gloves are sold by the dozen pairs.
 : The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.