Jomac® Extra Heavy Weight Terry Cloth Gloves, Loop Out, 2 ¹/₂" Cuff, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Jomac extra heavy weight terry cloth gloves resist heat up to 350 °F.
- Gauntlet cuff, ²¹/₂"
- Ambidextrous
- Loop-out terry cloth
- Unlined or lined style
They come in lined or unlined styles.
Ordering information: Gloves are sold by the dozen pairs.
Caution: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.
Specifications
- Catalog No:
- 80091-490
- 80091-488
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Cuff Style:
- Gauntlet
- Gauntlet