Order Entry
United States
Orders LinkContactUsLinkComponent
 

3-(3-Fluorophenyl)propionic acid ≥97.0%

3-(3-Fluorophenyl)propionic acid ≥97.0%
 
3-(3-Fluorophenyl)propionic acid ≥97.0%
3-(3-Fluorophenyl)propionic acid ≥97.0%

 

 
   
 

CAS Number: 458-45-7 MDL Number: MFCD01310821 Molecular Formula: C9H9FO2 Molecular Weight: 168.17 Purity/Analysis Method: 97.0% (GC,T) Form: Crystal Boiling point (°C): 119 Melting point (°C): 43

 :  3-(3-Fluorophenyl)propanoic acid
  •  :458-45-7
  •  :C₉H₉FO₂
  •  :MFCD01310821
  •  :168.17
  •  :119 °C
  •  :43 °C
 

 

  • Purity/Analysis Method:
    97.0% (GC,T)
Product Family Options

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.