Order Entry
United States
Orders LinkContactUsLinkComponent
Ammonium persulfate (APS) ≥99.0%

Ammonium persulfate (APS) ≥99.0%

Ammonium persulfate (APS) ≥99.0%
Supplier:  TCI America
Certificates
Ammonium persulfate (APS) ≥99.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 7727-54-0 MDL Number: MFCD00003390 Molecular Formula: H8N2O8S2 Molecular Weight: 228.19 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 120

Product Categories: Analytical Science, Reagents for Gel & Buffer Preparation, etc. (Reagents for Electrophoresis) Other Product Names / Synonyms: Ammonium Persulfate, APS

Synonyms:  Peroxidisulfuric acid diammonium salt, Ammonium persulfate, Peroxidisulphuric acid diammonium salt, Ammonium persulphate
  • CAS Number:7727-54-0
  • Formula:(NH₄)₂S₂O₈
  • MDL Number:MFCD00003390
  • Molecular Weight:228.2 g/mol
  • Melting Pt:120 °C
  • UN:1444
  • ADR:5.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    99.0% (T)
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.