Bacillus Subtilis (BGA) Spore Suspension, MilliporeSigma
Catalog # EM1.10649.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Ampoule
- Description:Bacillus Subtilis (BGA) Spore Suspension
- Size:15 mL
- Density:1 g/cm³
- Cat. No.:EM1.10649.0001
- Supplier no.:1.10649.0001
Specifications
About this item
For inhibitor tests.
- 8E+06 - 5E+07 CFU/mL viable spores in Bacillus subtilis DSM 618
- Passes performance test