Order Entry
United States
ContactUsLinkComponent
 
Anti-LY6G Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 1A8]
 
undefined
Anti-LY6G Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 1A8]

 

  • Catalog No:
  • 10050-542
  • 10051-598
  • Antigen Symbol:
  • LY6G
  • LY6G
  • Antigen Name:
  • Lymphocyte Antigen 6 Complex Locus G
  • Lymphocyte Antigen 6 Complex Locus G
  • Antigen Synonyms:
  • Gr1|Gr-1|Ly-6G
  • Gr1|Gr-1|Ly-6G
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • 1A8
  • 1A8
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 546644
  • 546644
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together