Anti-IgG Rabbit Polyclonal Antibody (CF™ 543)
Supplier: Biotium
Certificates
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Specifications
- Catalog No:
- 89411-510
- 89411-062
- Antigen Symbol:
- IgG
- IgG
- Antigen Name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 543
- CF® 543
- Reactivity:
- Goat
- Goat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Epitope:
- Heavy and Light chain
- Heavy and Light chain
Specifications
Frequently Bought Together