Anti-RORgt Mouse Monoclonal Antibody [clone: 4F3-3C8-2B7]
Catalog # 644902-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:RORgt
- Clonality:Monoclonal
- Clone:4F3-3C8-2B7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:19885
- Antigen Synonyms:RORC|Retinoid-Related Orphan Receptor gamma|Nuclear receptor RZR-gamma|ROR-g(t)|Nuclear receptor ROR-gamma|ROR gamma
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Human RORγ-GST recombinant protein
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:644902-BL
- Supplier no.:644902
Specifications
About this item
Purified anti-RORg [4F3-3C8-2B7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Weakly Cross-Reactive with Mouse; Apps: WB; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution: 1 to 4 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: RORgt
Clonality: Monoclonal
Clone: 4F3-3C8-2B7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Weakly Cross-Reactive with Mouse