Anti-p53 Mouse Monoclonal Antibody [clone: DO-7]
Supplier: Biolegend
Certificates
About this item
Purified anti-p53 [DO-7]; Isotype: Mouse IgG2b; Reactivity: Human; Apps: WB, ICC, ELISA; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 0.5 - 1.0 μg per ml antibody for each mini-gel. For immunocytochemistry, a concentration range of 2.0 - 5.0 μg/ml (Dilution 1:100 - 1:250) is recommended. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: DO-7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Specifications
- Catalog No:
- 645802-BL
- 645801-BL
- Antigen Symbol:
- p53
- p53
- Antigen Synonyms:
- Tumor protein p53 (TP53)
- Tumor protein p53 (TP53)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DO-7
- DO-7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7157
- 7157
- Isotype:
- IgG2b
- IgG2b
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Gel Shift:
- Yes
- Yes
- Immunogen:
- p53 N-terminal linear epitope aa 20-25
- p53 N-terminal linear epitope aa 20-25
- Form:
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- This antibody was purified by affinity chromatography.
- This antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together