Anti-Jagged 2 Mouse Monoclonal Antibody [clone: MHJ2-523]
Catalog # 346902-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Name:Jagged 2
- Clonality:Monoclonal
- Clone:MHJ2-523
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:3714
- Antigen Synonyms:Jag2|JAG2|Jagged-2
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Human Jagged2-transfected CHO cells
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:346902-BL
- Supplier no.:346902
Specifications
About this item
Purified anti-human Jagged 2 [MHJ2-523]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Jagged 2
Clonality: Monoclonal
Clone: MHJ2-523
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human