To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin A
- Clonality:Monoclonal
- Clone:E23.1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Bovine,Mouse
- Western Blot:Yes
- Size:25 μg
- Environmentally Preferable:
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:890
- Antigen Synonyms:CCNA1|CCN1|CCNA
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Cyclin A fusion protein
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:644001-BL
- Supplier no.:644001
Specifications
About this item
Purified anti-Cyclin A [E23.1]; Isotype: Mouse IgG2a; Reactivity: Human, Mouse, bovine; Apps: WB, ICFC; Size: 25 μg
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 0.5-1 μg per ml antibody dilution buffer. For intracellular flow cytometric staining, the suggested use of this reagent is 1 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Cyclin A
Clonality: Monoclonal
Clone: E23.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Bovine