Order Entry
United States
ContactUsLinkComponent
Anti-TNF-a Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: MAb11]
Anti-TNF-a Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: MAb11]
Supplier:  Biolegend
Certificates
Anti-TNF-a Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: MAb11]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 502926-BL
  • 502925-BL
  • Antigen Symbol:
  • TNF-a
  • TNF-a
  • Antigen Synonyms:
  • Differentiation inducing factor (DIF)|Necrosin|TNFSF2|TNFA|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
  • Differentiation inducing factor (DIF)|Necrosin|TNFSF2|TNFA|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Clone:
  • MAb11
  • MAb11
  • Reactivity:
  • Baboon, Pig, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
  • Baboon, Pig, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 7124
  • 7124
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • E. coli-expressed, recombinant human TNF-α
  • E. coli-expressed, recombinant human TNF-α
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
  • Storage Temperature:
  • The TNF-alpha antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The TNF-alpha antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together