PE anti-mouse/human Helios [22F6]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Human; Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. It is recommended that the reagent be titrated for optimal performance for each application.
Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: Helios
Clonality: Monoclonal
Clone: 22F6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Human
- Catalog No:
- 137206-BL
- 137216-BL
- Antigen Symbol:
- Helios
- Helios
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 22F6
- 22F6
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 22779, 22807
- 22779, 22807
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Immunogen:
- Helios peptide (aa 51-107)
- Helios peptide (aa 51-107)
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT