Anti-ACKR1 Rabbit Polyclonal Antibody
89514-012
: Abgent
- Antibody Type:Primary
- Antigen Name:Atypical Chemokine Receptor 1 (Duffy blood group)
- Antigen Symbol:ACKR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:2532
- Antigen Synonyms:CCBP1|CD234|GpFy|DARC|GPD|WBCQ1|FY|Dfy
- Amino Acid Number:1 to 30
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:35.553 kDa
- Sequence:PSTENSSQLDFEDVC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This DARC antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DARC.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89514-012
- Supplier no.:AP20261A
LEAF (Low Endotoxin, Azide-Free)
Western Blot: 1:1000
Type: Primary
Antigen: ACKR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human