Anti-BATF Rabbit Polyclonal Antibody
89519-582
: Abgent
- Antibody Type:Primary
- Antigen Name:Basic leucine zipper transcription factor, ATF-like
- Antigen Symbol:BATF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:10538
- Antigen Synonyms:SFA2|SFA-2|B-ATF|1
- Amino Acid Number:7 to 35
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:14.12 kDa
- Sequence:DSSDDVRRVQRREKC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This BATF antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 7-35 amino acids from the N-terminal region of human BATF.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89519-582
- Supplier no.:AP16614A
Western Blot: 1:1000
Type: Primary
Antigen: BATF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human