Anti-IL-10 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: JES5-16E3]
FITC anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES5-16E3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
- Catalog No:
- 505006-BL
- 505005-BL
- Antigen Symbol:
- IL-10
- IL-10
- Antigen Synonyms:
- B-TCGF|BTCGF|TGIF|Interleukin-10 (IL10)|IL10|CSIF
- B-TCGF|BTCGF|TGIF|Interleukin-10 (IL10)|IL10|CSIF
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- JES5-16E3
- JES5-16E3
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16153
- 16153
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- E. coli-expressed, recombinant mouse IL-10
- E. coli-expressed, recombinant mouse IL-10
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together