Specifications
- Antibody Type:Primary
- Antigen Symbol:IFN-g
- Clonality:Monoclonal
- Clone:B27
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Sooty mangabey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Size:100 tests
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:3458
- Antigen Synonyms:Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFNG|Interferon-γ|Immune interferon
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:Ambient RT
- Immunogen:E. coli-expressed recombinant human IFN-γ
- Purification:The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Cat. No.:506504-BL
- Supplier no.:506504
Specifications
About this item
FITC anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, African Green Monkey, Pigtailed Macaque, Sooty Mangabey; Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: B27
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;African Green Monkey;Pigtailed Macaque;Sooty Mangabey