Anti-ZRSR2 Rabbit Polyclonal Antibody
89513-548
: Abgent
- Antibody Type:Primary
- Antigen Name:Zinc Finger (CCCH type), RNA-binding motif And serine/arginine rich 2
- Antigen Symbol:ZRSR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:8233
- Antigen Synonyms:U2AF1RS2|U2AF1L2|U2AF1-RS2|ZC3H22|URP
- Amino Acid Number:453 to 482
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:58.045 kDa
- Sequence:CRSGNRDRTVQSPKSK
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This ZRSR2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 453-482 amino acids from the C-terminal region of human ZRSR2.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89513-548
- Supplier no.:AP20436B
Western Blot: 1:1000, Immunhistochemsitry: 1:25,
Type: Primary
Antigen: ZRSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human