About this item
APC anti-human/mouse CXCR7 [8F11-M16]; Isotype: Mouse IgG2b, κ; Reactivity: human, mouse; Apps: FC, ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: CXCR7
Clonality: Monoclonal
Clone: 8F11-M16
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: human;mouse
Specifications
- Catalog No:
- 331114-BL
- 331113-BL
- Antigen Symbol:
- CXCR7
- CXCR7
- Antigen Synonyms:
- RDC1|chemokine (C-X-C motif) receptor 7|G protein-coupled receptor
- RDC1|chemokine (C-X-C motif) receptor 7|G protein-coupled receptor
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 8F11-M16
- 8F11-M16
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 12778, 57007
- 12778, 57007
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Human CXCR7 transfectant
- Human CXCR7 transfectant
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT