Anti-IgG1, k isotype ctrl (ICFC) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: MOPC-21]
Alexa Fluor® 647 Mouse IgG1, κ isotype ctrl (ICFC) [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ICFC, IF; Size: 50 μg
Each lot of this mouse IgG1, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG1, k isotype ctrl (ICFC)
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
- Catalog No:
- 400136-BL
- 400155-BL
- Antigen Symbol:
- IgG1 k isotype ctrl (ICFC)
- IgG1 k isotype ctrl (ICFC)
- Antibody Type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Clone:
- MOPC-21
- MOPC-21
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Form:
- test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The immunoglobulin was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- The immunoglobulin was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Concentration:
- 0.50 mg/mL
- Storage Temperature:
- The mouse IgG1 kappa isotype control solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The mouse IgG1, κ isotype control solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together