Anti-CD66a/c/e Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: ASL-32]
Supplier: Biolegend
Certificates
About this item
Alexa Fluor® 488 anti-human CD66a/c/e [ASL-32]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD66a/c/e
Clonality: Monoclonal
Clone: ASL-32
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Specifications
- Catalog No:
- 342306-BL
- 342305-BL
- Antigen Symbol:
- CD66a/c/e
- CD66a/c/e
- Antigen Synonyms:
- CD66|CEACAM1/5/6|CD66ACE|NCA CEA|CEACAM|BGP
- CD66|CEACAM1/5/6|CD66ACE|NCA CEA|CEACAM|BGP
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Clone:
- ASL-32
- ASL-32
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 634
- 634
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together